Methyl 2-methoxypropionate, is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Methyl 2-methoxypropionate, is used as a pharmaceutical intermediate.
Solubility
Soluble in water.
Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
RUO – Research Use Only
H Kanai.; K Ishii. Hydrodimerization of methyl acrylate catalyzed by halogenotris (triphenylphosphine) cobalt. Bulletin of the Chemical Society of Japan. 1981, 54,(4), 1015-1018.
WA Bonner.; JA Zderic. The Stereochemistry of Raney Nickel Action. VI. Functional Group Hydrogenolyses in the α-Substituted α-Phenylpropionic Acid Series. Journal of the American Chemical Society. 1956, 78,(13), 3218-3221.