Thermo Scientific Chemicals

2-Nitroimidazole, 98%, Thermo Scientific Chemicals

Catalog number: L13546.ME
500 mg, Each
Thermo Scientific Chemicals

2-Nitroimidazole, 98%, Thermo Scientific Chemicals

Catalog number: L13546.ME
500 mg, Each
Quantity
Catalog number: L13546.ME
also known as L13546-ME
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
527-73-1
IUPAC Name
2-nitro-1H-imidazole
Molecular Formula
C3H3N3O2
InChI Key
YZEUHQHUFTYLPH-UHFFFAOYSA-N
SMILES
[O-][N+](=O)C1=NC=CN1
Assay (HPLC)
≥97.5%
Form
Crystals or powder or crystalline powder
Appearance (Color)
Pale yellow

Description

2-Nitroimidazole, is used as an antibiotic in pharma industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Nitroimidazole, is used as an antibiotic in pharma industry.

Solubility
It is slightly soluble in water. Soluble in acetone, butyl acetate, ethanol, ethyl acetate and methanol.

Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents, strong acids, acid chlorides, anhydrous acids.. Stable under recommended storage conditions.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text