2-Trifluoroacetyl-1,2,3,4-tetrahydroisoquinoline-7-sulfonyl chloride, 99%, Thermo Scientific Chemicals
2-Trifluoroacetyl-1,2,3,4-tetrahydroisoquinoline-7-sulfonyl chloride, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Trifluoroacetyl-1,2,3,4-tetrahydroisoquinoline-7-sulfonyl chloride, 99%, Thermo Scientific Chemicals

Have Questions?
Quantity:
5 g
Catalog number L13587.06
also known as L13587-06
Price (USD)/ Each
164.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
164.00
Add to cart
2-Trifluoroacetyl-1,2,3,4-tetrahydroisoquinoline-7-sulfonyl chloride, 99%, Thermo Scientific Chemicals
Catalog numberL13587.06
Price (USD)/ Each
164.00
-
Add to cart
Chemical Identifiers
CAS74291-57-9
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Melting Point (clear melt)100.0-106.0?C
FormCrystals or powder or crystalline powder
Assay (HPLC)≥98.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only