Thermo Scientific Chemicals

2,4-Dibromothiophene, 90+%, Thermo Scientific Chemicals

Catalog number: L13591.14
25 g, Each
Thermo Scientific Chemicals

2,4-Dibromothiophene, 90+%, Thermo Scientific Chemicals

Catalog number: L13591.14
25 g, Each
Quantity
Catalog number: L13591.14
also known as L13591-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3140-92-9
IUPAC Name
2,4-dibromothiophene
Molecular Formula
C4H2Br2S
InChI Key
WAQFYSJKIRRXLP-UHFFFAOYSA-N
SMILES
BrC1=CC(Br)=CS1
Form
Liquid
Appearance (Color)
Clear colorless to white to yellow to pale brown
Refractive Index
1.5920-1.6720 @ 20?C
Assay (GC)
≥90.0%

Description

2,4-Dibromothiophene is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used as an intermediate in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4-Dibromothiophene is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used as an intermediate in organic synthesis.

Solubility
It is insoluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text