Thermo Scientific Chemicals

2-(4-tert-Butylphenyl)ethanol, 96%, Thermo Scientific Chemicals

Catalog number: L13611.14
25 g, Each
Thermo Scientific Chemicals

2-(4-tert-Butylphenyl)ethanol, 96%, Thermo Scientific Chemicals

Catalog number: L13611.14
25 g, Each
Quantity
Catalog number: L13611.14
also known as L13611-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5406-86-0
IUPAC Name
2-(4-tert-butylphenyl)ethan-1-ol
Molecular Formula
C12H18O
InChI Key
NZGMMENPUKHODD-UHFFFAOYSA-N
SMILES
CC(C)(C)C1=CC=C(CCO)C=C1
Melting Point (clear melt)
29.0-35.0?C
Form
Fused solid or liquid as melt
Appearance (Color)
White or clear colorless
Assay (GC)
≥95.0%

Description

2-(4-tert-Butylphenyl)ethanol, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used as an intermediate in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(4-tert-Butylphenyl)ethanol, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used as an intermediate in organic synthesis.

Solubility
Insoluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text