2-Amino-5-n-propylsulfonylbenzimidazole, 98+%, Thermo Scientific Chemicals
2-Amino-5-n-propylsulfonylbenzimidazole, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Amino-5-n-propylsulfonylbenzimidazole, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
100 mg
Catalog number L13711.03
also known as L13711-03
Price (USD)/ Each
310.00
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
310.00
Add to cart
2-Amino-5-n-propylsulfonylbenzimidazole, 98+%, Thermo Scientific Chemicals
Catalog numberL13711.03
Price (USD)/ Each
310.00
-
Add to cart
Chemical Identifiers
CAS80983-34-2
IUPAC Name6-(propane-1-sulfonyl)-1H-1,3-benzodiazol-2-amine
Molecular FormulaC10H13N3O2S
InChI KeyWTPBIYSMFKUQKY-UHFFFAOYSA-N
SMILESCCCS(=O)(=O)C1=CC=C2N=C(N)NC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)≥98.0%
FormPowder
Melting Point220-228?C
Appearance (Color)White to cream to yellow or pale brown
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only