Petroleum ether 60/80, Thermo Scientific Chemicals
Petroleum ether 60/80, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Petroleum ether 60/80, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2500 mL
1000 mL
Catalog number L13734.0F
also known as L13734-0F
Price (USD)/ Each
57.65
Online exclusive
63.90 
Save 6.25 (10%)
-
Add to cart
Quantity:
2500 mL
Request bulk or custom format
Price (USD)/ Each
57.65
Online exclusive
63.90 
Save 6.25 (10%)
Add to cart
Petroleum ether 60/80, Thermo Scientific Chemicals
Catalog numberL13734.0F
Price (USD)/ Each
57.65
Online exclusive
63.90 
Save 6.25 (10%)
-
Add to cart
Chemical Identifiers
CAS8032-32-4
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Refractive Index1.3755-1.3865 @ 20?C
Identification (FTIR)Conforms
FormLiquid
It is mostly used by pharmaceutical companies and in the manufacturing process. It is a lightweight hydrocarbon used chiefly as a nonpolar solvent. The extract of aerial parts of ageratum conyzoids in petroleum ether plays an essential role for the isolation of sigmasterol and beta sitosterol. Crude petroleum spirit and methanol extracts also play an important role for the screening of antimicrobial activity of the stem bark, leaves and roots of Ficus thoningii.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is mostly used by pharmaceutical companies and in the manufacturing process. It is a lightweight hydrocarbon used chiefly as a nonpolar solvent. The extract of aerial parts of ageratum conyzoids in petroleum ether plays an essential role for the isolation of sigmasterol and beta sitosterol. Crude petroleum spirit and methanol extracts also play an important role for the screening of antimicrobial activity of the stem bark, leaves and roots of Ficus thoningii.

Solubility
Insoluble in water.

Notes
Flammable, may be moisture sensitive.
RUO – Research Use Only

General References:

  1. Kamboj, A.; Saluja, A. K. Isolation of Stigmasterol and beta sitosterol from petroleum ether extract of Aerial parts of Ageratum Conyzoides (Asteracea). Inter. J. Pharma. Pharma. Sci. 2011, 3 (1), 94­96.
  2. Ndukwe, I. G.; Bello, A. I.; Habila, J. D.; John, C. Phytochemical and antimicrobial screening of the crude petroleum spirit and methanol extracts of the stem bark, leaves and roots of Ficus thoningii (blume). Afr. J. Biotechnol. 2007, 6 (23), 2645-2649.