3-Hydroxy-2-methylbenzoic acid, 97%, Thermo Scientific Chemicals
3-Hydroxy-2-methylbenzoic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Hydroxy-2-methylbenzoic acid, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L13743.03
also known as L13743-03
Price (USD)/ Each
34.40
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
34.40
Add to cart
3-Hydroxy-2-methylbenzoic acid, 97%, Thermo Scientific Chemicals
Catalog numberL13743.03
Price (USD)/ Each
34.40
-
Add to cart
Chemical Identifiers
CAS603-80-5
IUPAC Name3-hydroxy-2-methylbenzoate
Molecular FormulaC8H7O3
InChI KeyRIERSGULWXEJKL-UHFFFAOYSA-M
SMILESCC1=C(O)C=CC=C1C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Cream to pale brown
Assay (Aqueous acid-base Titration)>96.0%
Formpowder
Assay (HPLC)>96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only