Thermo Scientific Chemicals

Pyrrole-2-carboxylic acid, 97%, Thermo Scientific Chemicals

Catalog number: L13756.14
25 g, Each
Thermo Scientific Chemicals

Pyrrole-2-carboxylic acid, 97%, Thermo Scientific Chemicals

Catalog number: L13756.14
25 g, Each
Quantity
Catalog number: L13756.14
also known as L13756-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
634-97-9
IUPAC Name
1H-pyrrole-2-carboxylic acid
Molecular Formula
C5H5NO2
InChI Key
WRHZVMBBRYBTKZ-UHFFFAOYSA-N
SMILES
OC(=O)C1=CC=CN1
Form
Crystals or powder or crystalline powder or lumps or fused solid or granules
Assay (Silylated GC)
≥96.0%
Assay (Aqueous acid-base Titration)
≥96.0 to ≤104.0%
Appearance (Color)
White to cream to pale brown or pink

Description

Pyrrole-2-carboxylic acid has demonstrated antiparasitic activity against Trypanosomes by selective proline racemase inhibition and has potent antifungal activity against Phytophthora. It also has antifungal properties.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pyrrole-2-carboxylic acid has demonstrated antiparasitic activity against Trypanosomes by selective proline racemase inhibition and has potent antifungal activity against Phytophthora. It also has antifungal properties.

Solubility
Soluble in methanol.

Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text