Thermo Scientific Chemicals

Isophorone diisocyanate, mixture of isomers, 98%, Thermo Scientific Chemicals

Catalog number: L13759.06
5 g, Each
Thermo Scientific Chemicals

Isophorone diisocyanate, mixture of isomers, 98%, Thermo Scientific Chemicals

Catalog number: L13759.06
5 g, Each
Quantity
Catalog number: L13759.06
also known as L13759-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4098-71-9
IUPAC Name
5-isocyanato-1-(isocyanatomethyl)-1,3,3-trimethylcyclohexane
Molecular Formula
C12H18N2O2
InChI Key
NIMLQBUJDJZYEJ-UHFFFAOYNA-N
SMILES
CC1(C)CC(CC(C)(CN=C=O)C1)N=C=O
Form
Liquid
Assay (GC)
≥97.5%
Appearance (Color)
Clear colorless to pale yellow
Identification (FTIR)
Conforms
Refractive Index
1.4825-1.4865 @ 20?C

Description

Isophorone diisocyanate is useful for the preparation of polyurethane plastics. It is also used to make polyurethane coatings and in enamel coatings, which are resistant to abrasion and degradation from ultraviolet light. Further, it is used as coatings for automotive, flooring and roofing as well as paint applied to aircraft.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Isophorone diisocyanate is useful for the preparation of polyurethane plastics. It is also used to make polyurethane coatings and in enamel coatings, which are resistant to abrasion and degradation from ultraviolet light. Further, it is used as coatings for automotive, flooring and roofing as well as paint applied to aircraft.

Solubility
Miscible with esters, ketones, ethers, and aromatic and aliphatic hydrocarbons.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents, strong acids and alcohols.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text