Thermo Scientific Chemicals

Perfluorotetradecanoic acid, 96%, Thermo Scientific Chemicals

Catalog number: L13796.03
1 g, Each
Thermo Scientific Chemicals

Perfluorotetradecanoic acid, 96%, Thermo Scientific Chemicals

Catalog number: L13796.03
1 g, Each
Quantity
Catalog number: L13796.03
also known as L13796-03
Price (USD)
Have Questions?

Chemical Identifiers

CAS
376-06-7
IUPAC Name
heptacosafluorotetradecanoic acid
Molecular Formula
C14HF27O2
InChI Key
RUDINRUXCKIXAJ-UHFFFAOYSA-N
SMILES
OC(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
Appearance (Color)
White
Assay (Aqueous acid-base Titration)
≥95.0 to ≤105.0%
Identification (FTIR)
Conforms
Form
Powder and/or Lumps
Melting Point (clear melt)
127-140?C

Description

Perfluorotetradecanoic acid is used for GC (Gas Chromatography) and LC (Liquid Chromatography) analysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Perfluorotetradecanoic acid is used for GC (Gas Chromatography) and LC (Liquid Chromatography) analysis.

Solubility
It is insoluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text