Perfluorotetradecanoic acid, 96%, Thermo Scientific Chemicals
Perfluorotetradecanoic acid, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Perfluorotetradecanoic acid, 96%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number L13796.06
also known as L13796-06
Price (USD)/ Each
65.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
76.50 
Save 10.85 (14%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
65.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
76.50 
Save 10.85 (14%)
Add to cart
Perfluorotetradecanoic acid, 96%, Thermo Scientific Chemicals
Catalog numberL13796.06
Price (USD)/ Each
65.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
76.50 
Save 10.85 (14%)
-
Add to cart
Chemical Identifiers
CAS376-06-7
IUPAC Nameheptacosafluorotetradecanoic acid
Molecular FormulaC14HF27O2
InChI KeyRUDINRUXCKIXAJ-UHFFFAOYSA-N
SMILESOC(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (Aqueous acid-base Titration)≥95.0 to ≤105.0%
Identification (FTIR)Conforms
FormPowder and/or Lumps
Melting Point (clear melt)127-140?C
Perfluorotetradecanoic acid is used for GC (Gas Chromatography) and LC (Liquid Chromatography) analysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Perfluorotetradecanoic acid is used for GC (Gas Chromatography) and LC (Liquid Chromatography) analysis.

Solubility
It is insoluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
RUO – Research Use Only

General References:

  1. HJ Lehmler.; PM Bummer. Mixing of perfluorinated carboxylic acids with dipalmitoylphosphatidylcholine. Biochimica et Biophysica Acta . 2004, 1664,(2), 141-149.
  2. HK Park.; TH Ha.; K Kim. Gelation of octadecyltrimethoxysilane at the air/water interface: Effect ofperfluorotetradecanoic acid and divalent cadmium ions. Langmuir. 2004, 20,(12), 4851-4858.