Thermo Scientific Chemicals

4-Bromo-3-chlorophenol, 98%, Thermo Scientific Chemicals

Catalog number: L13801.06
5 g, Each
Thermo Scientific Chemicals

4-Bromo-3-chlorophenol, 98%, Thermo Scientific Chemicals

Catalog number: L13801.06
5 g, Each
Quantity
Catalog number: L13801.06
also known as L13801-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13631-21-5
IUPAC Name
4-bromo-3-chlorophenol
Molecular Formula
C6H4BrClO
InChI Key
FQEYHIPPYOSPLF-UHFFFAOYSA-N
SMILES
OC1=CC(Cl)=C(Br)C=C1
Appearance (Color)
White to pale cream to cream to yellow to pale brown
Assay (GC)
≥97.5%
Form
Crystals or powder or crystalline powder or fused solid
Melting Point (clear melt)
62.0-69.0?C

Description

4-Bromo-3-chlorophenol is used as a precursor in organic synthesis to prepare 4-(benzyloxy)-l-bromo-2-chlorobenzene by reaction with benzyl bromide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromo-3-chlorophenol is used as a precursor in organic synthesis to prepare 4-(benzyloxy)-l-bromo-2-chlorobenzene by reaction with benzyl bromide.

Solubility
Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text