Thermo Scientific Chemicals

Quinoline-3-carboxaldehyde, 98+%, Thermo Scientific Chemicals

Catalog number: L13815.MD
250 mg, Each
Thermo Scientific Chemicals

Quinoline-3-carboxaldehyde, 98+%, Thermo Scientific Chemicals

Catalog number: L13815.MD
250 mg, Each
Quantity
Catalog number: L13815.MD
also known as L13815-MD
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13669-42-6
IUPAC Name
quinoline-3-carbaldehyde
Molecular Formula
C10H7NO
InChI Key
RYGIHSLRMNXWCN-UHFFFAOYSA-N
SMILES
O=CC1=CN=C2C=CC=CC2=C1
Free acid (titration)
≤1% (non-U.S. sourced material)
Appearance (Color)
White to dark cream to cream to yellow to brown
Form
Crystals or powder or crystalline powder or fused solid
Comment
Material sourced in the U.S. and in other countries
Assay (GC)
≥98.0%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text