Thermo Scientific Chemicals

N-Methylaniline, 98%, Thermo Scientific Chemicals

Catalog number: L13873.0E
2500 g, Each
Thermo Scientific Chemicals

N-Methylaniline, 98%, Thermo Scientific Chemicals

Catalog number: L13873.0E
2500 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: L13873.0E
also known as L13873-0E
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
100-61-8
IUPAC Name
N-methylaniline
Molecular Formula
C7H9N
InChI Key
AFBPFSWMIHJQDM-UHFFFAOYSA-N
SMILES
CNC1=CC=CC=C1
Appearance (Color)
Clear colorless to yellow
Assay (GC)
>97.5%
Form
Liquid
Refractive Index
1.5685-1.5725 @ 20?C

Description

N-Methylaniline is used in the preparation of N,N'-dimethylsulfinyldianilin using triethylamine and thionyl chloride as reagents. It is also acts as a latent and coupling solvent. Further, it serves as an intermediate for dyes and agrochemicals. In addition to this, it is utilized as an antiknock agent, which plays an essential role to increase the octane number.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Methylaniline is used in the preparation of N,N′-dimethylsulfinyldianilin using triethylamine and thionyl chloride as reagents. It is also acts as a latent and coupling solvent. Further, it serves as an intermediate for dyes and agrochemicals. In addition to this, it is utilized as an antiknock agent, which plays an essential role to increase the octane number.

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agents, acids, acid anhydrides, acid chlorides and carbon dioxide.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text