Imidazole-2-carboxaldehyde is a novel protein tyrosine phosphatase 1B (PTP1B) inhibitor with an important application to treat type-2 diabetes. It is used in the preparation of tridentate Schiff-base carboxylate-containing ligands by undergoing condensation reaction with amino acids beta-alanine and 2-aminobenzoic acid. It is also involved in the study of the imidazole-directed allylation of aldimines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Imidazole-2-carboxaldehyde is a novel protein tyrosine phosphatase 1B (PTP1B) inhibitor with an important application to treat type-2 diabetes. It is used in the preparation of tridentate Schiff-base carboxylate-containing ligands by undergoing condensation reaction with amino acids beta-alanine and 2-aminobenzoic acid. It is also involved in the study of the imidazole-directed allylation of aldimines.
Solubility
Soluble in water.
Notes
Air sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
RUO – Research Use Only
Joseyphus, R. S.; Shiju, C.; Joseph, J.; Dhanaraj, C. J.; Arish, D. Synthesis and characterization of metal complexes of Schiff base ligand derived from imidazole-2-carboxaldehyde and 4-aminoantipyrine. Spectrochim. Acta, Part A 2014, 133, 149-155.
Tinel, L.; Dumas, S.; George, C. A time-resolved study of the multiphase chemistry of excited carbonyls: Imidazole-2-carboxaldehyde and halides. C. R. Chim. 2014, 17 (7), 801-807.