3-Diethylaminopropylamine is used as a solvent, extraction agents and organic synthesis intermediates, dyes, pigments, surface active agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Diethylaminopropylamine is used as a solvent, extraction agents and organic synthesis intermediates, dyes, pigments, surface active agent.
Solubility
Fully miscible in water.
Notes
Air sensitive. Store under inert gas. Store away from oxidizing agents, air.
RUO – Research Use Only
General References:
- John Burkus. Tertiary Amine Catalysis of the Reaction of Phenyl Isocyanate with Alcohols. J. Org. Chem. 1961, 26, (3), 779-782.
- Kaushal Rege; Asif Ladiwala; Nihal Tugcu; Curt M. Breneman; Steven M. Cramer. Parallel screening of selective and high-affinity displacers for proteins in ion-exchange systems. Journal of Chromatography A. 2004, 1033, (1), 19-28.