Thermo Scientific Chemicals

Tetrahydrothiophene, 98%, Thermo Scientific Chemicals

Catalog number: L13908.22
100 g, Each
Thermo Scientific Chemicals

Tetrahydrothiophene, 98%, Thermo Scientific Chemicals

Catalog number: L13908.22
100 g, Each
Quantity
Catalog number: L13908.22
also known as L13908-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
110-01-0
IUPAC Name
thiolane
Molecular Formula
C4H8S
InChI Key
RAOIDOHSFRTOEL-UHFFFAOYSA-N
SMILES
C1CCSC1
Appearance (Color)
Clear colorless
Assay (GC)
≥97.5%
Form
Liquid
Refractive Index
1.5030-1.5070 @ 20°C
Identification (FTIR)
Conforms

Description

Tetrahydrothiophene is used as pharmaceutical and agrochemical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetrahydrothiophene is used as pharmaceutical and agrochemical intermediates.

Solubility
Immiscible in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text