Thermo Scientific Chemicals

Isopentyl nitrite, 97%, stab. with 0.2% anhyd. sodium carbonate, Thermo Scientific Chemicals

Catalog number: L13909.AP
500 mL, Each
Thermo Scientific Chemicals

Isopentyl nitrite, 97%, stab. with 0.2% anhyd. sodium carbonate, Thermo Scientific Chemicals

Catalog number: L13909.AP
500 mL, Each
Quantity
Catalog number: L13909.AP
also known as L13909-AP
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
110-46-3
IUPAC Name
3-methylbutyl nitrite
Molecular Formula
C5H11NO2
InChI Key
OWFXIOWLTKNBAP-UHFFFAOYSA-N
SMILES
CC(C)CCON=O
Identification (FTIR)
Conforms
Refractive Index
1.3845-1.3895 @20?C
Appearance (Color)
Clear colorless to yellow or pale orange
Assay (GC)
≥96.0%
Form
Liquid

Description

Isopentyl nitrite is used in the conversion of 2,6-disubstituted phenols to diphenoquinones and aminophenols to quinone diazides. It acts as an oxidant and nitrosating agent. It finds application in aromatic arylation, deamination of arylamines using tetrahydrofuran as a hydrogen donor. It is also useful for the preparation of benzyne using anthranilic acid. It is an antihypertensive and vasodilator to treat heart diseases such as angina and an antidote for cyanide poisoning. Further, it is used as a solvent and cleaning agent for a printed circuit board. In addition to this, it is considered to be a replacement for freon.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Miscible with methanol, chloroform and ether. Slightly miscible with water.

Notes
Air, moisture and light sensitive. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents and alcohol.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text