Thermo Scientific Chemicals

Dowex™ 50WX4 100-200 (H), Thermo Scientific Chemicals

Catalog number: L13917.0B
1000 g, Each
Thermo Scientific Chemicals

Dowex™ 50WX4 100-200 (H), Thermo Scientific Chemicals

Catalog number: L13917.0B
1000 g, Each
Quantity
Catalog number: L13917.0B
also known as L13917-0B
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
11113-61-4
MDL Number
MFCD00132725
Form
Powder or beads
Comment
Polystyrene crosslinked with 4% divinylbenzene
Moisture holding capacity
64-72%
Appearance (Color)
Pale orange to pale brown
Comment
Ionic form: H+

Description

Dowex™ 50WX4 100-200 (H) is used for the conversion of D,L-glyceraldehyde 3-phosphate diethyl acetal (barium salt) on treatment with boiling water to free aldehyde D,L-glyceraldehyde 3-phosphate. It is also used to investigate the effect of particle size on drug loading and release profiles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dowex|r 50WX4 100-200 (H) is used for the conversion of D,L-glyceraldehyde 3-phosphate diethyl acetal (barium salt) on treatment with boiling water to free aldehyde D,L-glyceraldehyde 3-phosphate. It is also used to investigate the effect of particle size on drug loading and release profiles.

Solubility
Insoluble in water.

Notes
Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text