Thermo Scientific Chemicals

L-2-Aminoadipic acid, 98%, Thermo Scientific Chemicals

Catalog number: L13924.06
5 g, Each
Thermo Scientific Chemicals

L-2-Aminoadipic acid, 98%, Thermo Scientific Chemicals

Catalog number: L13924.06
5 g, Each
Quantity
Catalog number: L13924.06
also known as L13924-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1118-90-7
IUPAC Name
(2S)-2-aminohexanedioic acid
Molecular Formula
C6H11NO4
InChI Key
OYIFNHCXNCRBQI-BYPYZUCNSA-N
SMILES
N[C@@H](CCCC(O)=O)C(O)=O
Optical Rotation
+24° ± 2° (c=2 in 5N HCl, non-U.S. specification)
Appearance (Color)
White
Form
Powder
Acid-base back titration
≥97.5 to ≤102.5% (non-U.S. specification)
Assay (Silylated GC)
≥97.5% (non-U.S. specification)

Description

L-2-Aminoadipic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-2-Aminoadipic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

Solubility
Soluble in 1N HCl (50 mg/ml) and water (slightly).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text