Appearance (Color)White
FormCrystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5% (non-U.S. specification)
Assay from Suppliers CofA≥97.5% (U.S. specification)
CommentSpecification differs for U.S. and non-U.S. material where indicated
View more