Thermo Scientific Chemicals

N-Phenyldiethanolamine, 97%, Thermo Scientific Chemicals

Catalog number: L13931.0E
2500 g, Each
Thermo Scientific Chemicals

N-Phenyldiethanolamine, 97%, Thermo Scientific Chemicals

Catalog number: L13931.0E
2500 g, Each
Quantity
Catalog number: L13931.0E
also known as L13931-0E
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
120-07-0
IUPAC Name
2-[(2-hydroxyethyl)(phenyl)amino]ethan-1-ol
Molecular Formula
C10H15NO2
InChI Key
OJPDDQSCZGTACX-UHFFFAOYSA-N
SMILES
OCCN(CCO)C1=CC=CC=C1
Melting Point (clear melt)
54.0-62.0°C
Appearance (Color)
White to cream to dark yellow to pale brown or blue to purple
Form
Crystals or powder or crystalline powder or lumps or chunks or fused solid
Assay (GC)
≥96.0%
Identification (FTIR)
Conforms

Description

N-Phenyldiethanolamine is used as phenylephrine intermediates, used as dye intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Phenyldiethanolamine is used as phenylephrine intermediates, used as dye intermediates.

Solubility
Soluble in water 3g/L @ 20˚C.

Notes
Air sensitive & hygroscopic. Store under inert gas. Store away from oxidizing agents, air, water/ moisture.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text