N-Phenyldiethanolamine is used as phenylephrine intermediates, used as dye intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N-Phenyldiethanolamine is used as phenylephrine intermediates, used as dye intermediates.
Solubility
Soluble in water 3g/L @ 20˚C.
Notes
Air sensitive & hygroscopic. Store under inert gas. Store away from oxidizing agents, air, water/ moisture.
RUO – Research Use Only
Paul B. Hodgson; Fabrice H. Salingue. The preparation of a stable 2-pyridylboronate and its reactivity in the Suzuki-Miyaura cross-coupling reaction. Tetrahedron Letters. 2004, 45, (4), 685-687.
Yoshiaki Shirae; Takashi Mino; Tae Hasegawa; Masami Sakamoto;Tsutomu Fujita. Transesterification of various alcohols with vinyl acetate under mild conditions catalyzed by diethylzinc using N-substituted diethanolamine as a ligand. Tetrahedron Letters. 1987, 46, (35), 5877-5879.