Thermo Scientific Chemicals

2,5-Norbornadiene, 97%, stab with 250 ppm BHT, Thermo Scientific Chemicals

Catalog number: L13935.AC
25 mL, Each
Thermo Scientific Chemicals

2,5-Norbornadiene, 97%, stab with 250 ppm BHT, Thermo Scientific Chemicals

Catalog number: L13935.AC
25 mL, Each
Quantity
Catalog number: L13935.AC
also known as L13935-AC
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
121-46-0
IUPAC Name
bicyclo[2.2.1]hepta-2,5-diene
Molecular Formula
C7H8
InChI Key
SJYNFBVQFBRSIB-UHFFFAOYSA-N
SMILES
C1C2C=CC1C=C2
Assay (GC)
≥96.0% (non-U.S. sourced material)
Comment
Material sourced in the U.S. and in other countries
Form
Liquid
Assay from Supplier's CofA
≥96.0% (U.S. sourced material)
Refractive Index
1.4685-1.4735 @ 20°C (non-U.S. sourced material)

Description

2,5-Norbornadiene is used as an intermediate in prostaglandin synthesis and shows anti-ethylene effect on plants. It acts as a starting material for the synthesis of diamantane and as an acetylene transfer agent for instance, in reaction with 3,6-di-2-pyridyl-1,2,4,5-tetrazine. It is also a useful dienophile in Diels-Alder reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,5-Norbornadiene is used as an intermediate in prostaglandin synthesis and shows anti-ethylene effect on plants. It acts as a starting material for the synthesis of diamantane and as an acetylene transfer agent for instance, in reaction with 3,6-di-2-pyridyl-1,2,4,5-tetrazine. It is also a useful dienophile in Diels-Alder reactions.

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agents, strong acids and strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text