Thermo Scientific Chemicals

Urea hydrogen peroxide adduct, 97%, Thermo Scientific Chemicals

Catalog number: L13940.36
500 g, Each
Thermo Scientific Chemicals

Urea hydrogen peroxide adduct, 97%, Thermo Scientific Chemicals

Catalog number: L13940.36
500 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: L13940.36
also known as L13940-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
124-43-6
IUPAC Name
peroxol; urea
Molecular Formula
CH6N2O3
InChI Key
AQLJVWUFPCUVLO-UHFFFAOYSA-N
SMILES
OO.NC(N)=O
Melting Point (clear melt)
87-93?C
Form
Crystals or crystalline powder or tables
Identification (FTIR)
Conforms
Comment
Available oxygen 16-18%
Appearance (Color)
White

Description

Urea hydrogen peroxide is an antiseptic, disinfectant and bleaching agent used in pharmaceuticals and cosmetics respectively. It is used in the whitening of teeth and relieves minor inflammation of gums, oral mucosal surfaces and lips. It finds application in the preparation of plastics, blue print development and starch modification. In addition to this, it is used as a source of hydrogen peroxide easily handled in the laboratory.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Urea hydrogen peroxide is an antiseptic, disinfectant and bleaching agent used in pharmaceuticals and cosmetics respectively. It is used in the whitening of teeth and relieves minor inflammation of gums, oral mucosal surfaces and lips. It finds application in the preparation of plastics, blue print development and starch modification. In addition to this, it is used as a source of hydrogen peroxide easily handled in the laboratory.

Solubility
Soluble in water.

Notes
Incompatible with acetic anhydride, aniline and carboxylic acids.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text