Thermo Scientific Chemicals

AmberChrom 50Wx2 200-400 (H), Thermo Scientific Chemicals

Catalog number: L13945.0B
1000 g, Each
Thermo Scientific Chemicals

AmberChrom 50Wx2 200-400 (H), Thermo Scientific Chemicals

Catalog number: L13945.0B
1000 g, Each
Quantity
Catalog number: L13945.0B
also known as L13945-0B
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
12612-37-2
MDL Number
MFCD00132722
Capacity
≥0.6 eq/L by wetted bed volume
Comment
Polystyrene cross-linked with 2% divinylbenzene matrix.
Comment
Sulphonic acid functional group as main active group.
Moisture holding capacity
74-82%
pH
range: 0-14

Description

Dowex™ 50WX2 200-400 (H) is used as a catalyst for selective monotetrahydropyranylation of symmetrical diols and for selective monoesterification of dicarboxylic acids. It is also used to remove calcium from aqueous samples.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dowex™ 50WX2 200-400 (H) is used as a catalyst for selective monotetrahydropyranylation of symmetrical diols and for selective monoesterification of dicarboxylic acids. It is also used to remove calcium from aqueous samples.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text