(R)-(-)-2-Butanol is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
(R)-(-)-2-Butanol is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.
Solubility
Partly miscible in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.
RUO – Research Use Only
Van T. Pham; Robert S. Phillips; Lars G. Ljungdahl. Temperature-dependent enantiospecificity of secondary alcohol dehydrogenase from Thermoanaerobacter ethanolicus. J. Am. Chem. Soc. 1989, 111, (5), 1935-1936.
Feng Wang and Prasad L. Polavarapu. Vibrational Circular Dichroism: Predominant Conformations and Intermolecular Interactions in (R)-(-)-2-Butanol. J. Phys. Chem. A. 2000, 104, (46), 10683-10687.