Thermo Scientific Chemicals

(R)-(-)-2-Butanol, 98+%, Thermo Scientific Chemicals

Catalog number: L13983.06
5 g, Each
Thermo Scientific Chemicals

(R)-(-)-2-Butanol, 98+%, Thermo Scientific Chemicals

Catalog number: L13983.06
5 g, Each
Quantity
Catalog number: L13983.06
also known as L13983-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
14898-79-4
IUPAC Name
(2R)-butan-2-ol
Molecular Formula
C4H10O
InChI Key
BTANRVKWQNVYAZ-SCSAIBSYSA-N
SMILES
CC[C@@H](C)O
Appearance (Color)
Clear colorless
Optical Rotation
-14.0? ? 2.0? (neat)
Assay (GC)
≥98.0%
Refractive Index
1.3960-1.4000 @ 20?C
Form
Liquid

Description

(R)-(-)-2-Butanol is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-(-)-2-Butanol is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Partly miscible in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text