Thermo Scientific Chemicals

L-(+)-2-Aminobutyric acid, 98+%, Thermo Scientific Chemicals

Catalog number: L13984.14
25 g, Each
Thermo Scientific Chemicals

L-(+)-2-Aminobutyric acid, 98+%, Thermo Scientific Chemicals

Catalog number: L13984.14
25 g, Each
Quantity
Catalog number: L13984.14
also known as L13984-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1492-24-6
IUPAC Name
2-aminobutanoic acid
Molecular Formula
C4H9NO2
InChI Key
QWCKQJZIFLGMSD-UHFFFAOYNA-N
SMILES
CCC(N)C(O)=O
Assay (Non-aqueous acid-base Titration)
≥98.0 to ≤102.0%
Form
Crystals or powder or crystalline powder or flakes
Optical Rotation
20.8 ± 1.0° (c=4.8 in 5N HCl)
Appearance (Color)
White to pale cream to pale brown

Description

L-(+)-2-Aminobutyric acid is used in the biosynthesis of nonribosomal peptides. It acts as a receptor antagonist. It is also used as a chiral reagent. Further, it is used in the determination of substrate of glutamyl cysteine acid synthase. In addition to this, it is also utilized as a drug intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-(+)-2-Aminobutyric acid is used in the biosynthesis of nonribosomal peptides. It acts as a receptor antagonist. It is also used as a chiral reagent. Further, it is used in the determination of substrate of glutamyl cysteine acid synthase. In addition to this, it is also utilized as a drug intermediate.

Solubility
Soluble in water and acetic acid. Slightly soluble in alcohol and ether.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text