Thermo Scientific Chemicals

(1S)-(+)-Menthyl acetate, 99%, Thermo Scientific Chemicals

Catalog number: L13987.MD
250 mg, Each
Thermo Scientific Chemicals

(1S)-(+)-Menthyl acetate, 99%, Thermo Scientific Chemicals

Catalog number: L13987.MD
250 mg, Each
Quantity
Catalog number: L13987.MD
also known as L13987-MD
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5157-89-1
IUPAC Name
5-methyl-2-(propan-2-yl)cyclohexyl acetate
Molecular Formula
C12H22O2
InChI Key
XHXUANMFYXWVNG-UHFFFAOYNA-N
SMILES
CC(C)C1CCC(C)CC1OC(C)=O
Assay (GC)
≥98.0%
Refractive Index
1.4450-1.4470 @ 20?C
Form
Liquid
Appearance (Color)
Clear colorless to pale yellow
Optical Rotation
+82? ±2? (c=2 in Ethanol)

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text