Thermo Scientific™

Silica gel 60, 0.036-0.071mm (215-400 mesh), Thermo Scientific Chemicals

Catalog number: L14004.0B
1000 g, Each
Thermo Scientific™

Silica gel 60, 0.036-0.071mm (215-400 mesh), Thermo Scientific Chemicals

Catalog number: L14004.0B
1000 g, Each
Quantity
Catalog number: L14004.0B
also known as L14004-0B
Price (EUR)
Quantity
-

Chemical Identifiers

CAS
7631-86-9
IUPAC Name
silanedione
Molecular Formula
O2Si
InChI Key
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
SMILES
O=[Si]=O
Comment
pH (5% aqueous suspension):6.5 - 7.5
Particle Size
≤ 10.0 wt% @ > 71?m
Appearance (Color)
White
Form
powder
Particle Size
≤ 10.0 wt% @ < 36?m

Description

Silica gel is used in column chromatography without applied pressure (gravity pulled). It is used as an adsorbent in various applications and in removing impurities. It is used as stationary base in column chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Silica gel is used in column chromatography without applied pressure (gravity pulled). It is used as an adsorbent in various applications and in removing impurities. It is used as stationary base in column chromatography.

Solubility
Soluble in hydrofluoric acid. Insoluble in water.

Notes
Keep the container tightly sealed. Store in cool, dry conditions in a well sealed container. Incompatible with hydrogen fluoride.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text