Thermo Scientific Chemicals

Acetyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: L14013.AC
25 mL, Each
Thermo Scientific Chemicals

Acetyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: L14013.AC
25 mL, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: L14013.AC
also known as L14013-AC
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
75-36-5
IUPAC Name
acetyl chloride
Molecular Formula
C2H3ClO
InChI Key
WETWJCDKMRHUPV-UHFFFAOYSA-N
SMILES
CC(Cl)=O
Assay (GC)
≥97.5%
Form
Liquid
Identification (FTIR)
Conforms
Appearance (Color)
Clear colorless to pale pink
Refractive Index
1.3875-1.3915 @ 20?C

Description

Acetyl chloride acts as a reagent for the preparation of esters and amides of acetic acid. It is also useful as an important reactant in Friedel-Crafts reactions as well as in the introduction of an acetyl group. It serves as a starting material in the production of pharmaceutical, new plating complexing agent, acylation agent and synthetic organic intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Acetyl chloride acts as a reagent for the preparation of esters and amides of acetic acid. It is also useful as an important reactant in Friedel-Crafts reactions as well as in the introduction of an acetyl group. It serves as a starting material in the production of pharmaceutical, new plating complexing agent, acylation agent and synthetic organic intermediates.

Solubility
Miscible with acetone, chloroform, glacial acetic acid, petroleum ether, ether and benzene.

Notes
Moisture sensitive. Incompatible with water, alcohols, oxidizing agents and strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text