Adenosine-5'-diphosphate disodium salt is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Adenosine-5′-diphosphate disodium salt is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature is 20°C. Store under inert gas. Moisture sensitive. It is hygroscopic in nature. Incompatible with strong oxidizing agents.
RUO – Research Use Only
Yasukazu Yoshida.; Satoshi Furuta.; Etsuo Niki. Effects of metal chelating agents on the oxidation of lipids induced by copper and iron. Biochimica et Biophysica Acta (BBA) - Lipids and Lipid Metabolism. 1993, 1210 (1), 81-88.