Thermo Scientific Chemicals

Adenosine-5'-diphosphate disodium salt, 96% (dry wt.), water 15% max.

Catalog number: L14029.MD
250 mg, Each
Thermo Scientific Chemicals

Adenosine-5'-diphosphate disodium salt, 96% (dry wt.), water 15% max.

Catalog number: L14029.MD
250 mg, Each
Quantity
Catalog number: L14029.MD
also known as L14029-MD
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
16178-48-6
IUPAC Name
disodium hydrogen ({[(2R,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphonato}oxy)phosphonate
Molecular Formula
C10H13N5Na2O10P2
InChI Key
ORKSTPSQHZNDSC-WCYUCLFNNA-L
SMILES
[Na+].[Na+].NC1=C2N=CN([C@@H]3O[C@H](COP([O-])(=O)OP(O)([O-])=O)[C@@H](O)[C@H]3O)C2=NC=N1
Appearance (Color)
White to pale cream
Assay from Supplier's CofA
≥95% (dry weight basis)
Optical Rotation
-31.5? ± 2.0? (c=0.5 in 0.5M Na2HPO4) (dry wt. basis)
Water Content (Karl Fischer Titration)
≤15%
Form
Powder

Description

Adenosine-5'-diphosphate disodium salt is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Adenosine-5′-diphosphate disodium salt is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature is 20°C. Store under inert gas. Moisture sensitive. It is hygroscopic in nature. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text