Thermo Scientific Chemicals

Tetrafluoroboric acid, ca 50% w/w aq. soln., Thermo Scientific Chemicals

Catalog number: L14037.36
500 g, Each
Thermo Scientific Chemicals

Tetrafluoroboric acid, ca 50% w/w aq. soln., Thermo Scientific Chemicals

Catalog number: L14037.36
500 g, Each
Quantity
Catalog number: L14037.36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
16872-11-0
IUPAC Name
hydrogen tetrafluoroboranuide
Molecular Formula
BF4H
InChI Key
ODGCEQLVLXJUCC-UHFFFAOYSA-O
SMILES
[H+].F[B-](F)(F)F
Impurity content
Fluorosilicic acid: ≤200ppm (U.K. specification)
Impurity content
Sulfate: ≤200ppm (U.K. specification)
Heavy Metals
≤500ppm (as lead) (U.S. and U.K. specification)
Appearance (Color)
Clear colorless to pale yellow
Chloride content
≤50ppm (U.K. specification)

Description

Tetrafluoroboric acid is used in conjunction with various methods of cleaning and to treat the surface of substrates. It is also used as a radiotracer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetrafluoroboric acid is used in conjunction with various methods of cleaning and to treat the surface of substrates. It is also used as a radiotracer.

Solubility
Miscible with water.

Notes
Incompatible with strong bases, oxidizing agents and acids.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text