Thermo Scientific Chemicals

3-(2-Aminoethylamino)propyltrimethoxysilane, 96%, Thermo Scientific Chemicals

Catalog number: L14043.14
25 g, Each
Thermo Scientific Chemicals

3-(2-Aminoethylamino)propyltrimethoxysilane, 96%, Thermo Scientific Chemicals

Catalog number: L14043.14
25 g, Each
Quantity
Catalog number: L14043.14
also known as L14043-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1760-24-3
IUPAC Name
hydrogen (2-aminoethyl)[3-(trimethoxysilyl)propyl]amine chloride
Molecular Formula
C8H23ClN2O3Si
InChI Key
SZBIXNNLIKDOEJ-UHFFFAOYSA-N
SMILES
[H+].[Cl-].CO[Si](CCCNCCN)(OC)OC
Appearance (Color)
Clear colorless to yellow
Form
Liquid
Assay (GC)
≥95.0%
Identification (FTIR)
Conforms
Refractive Index
1.4415-1.4475 @ 20?C

Description

3-(2-Aminoethylamino) propyltrimethoxysilane acts as a silane coupling agent. It is useful for chemical research. It is also employed as an adhesives and sealant chemical. Further, it is used as a paint additive and coating additive. In addition to this, it plays an important role in the production of petroleum.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-(2-Aminoethylamino) propyltrimethoxysilane acts as a silane coupling agent. It is useful for chemical research. It is also employed as an adhesives and sealant chemical. Further, it is used as a paint additive and coating additive. In addition to this, it plays an important role in the production of petroleum.

Solubility
Miscible with toluene.

Notes
Air and moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents and water.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text