Thermo Scientific Chemicals

Adenosine-5'-monophosphoric acid, 99% (dry wt.), water <6%

Catalog number: L14051.03
1 g, Each
Thermo Scientific Chemicals

Adenosine-5'-monophosphoric acid, 99% (dry wt.), water <6%

Catalog number: L14051.03
1 g, Each
Quantity
Catalog number: L14051.03
also known as L14051-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
61-19-8
IUPAC Name
{[(2R,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy}phosphonic acid
Molecular Formula
C10H14N5O7P
InChI Key
UDMBCSSLTHHNCD-YPLCUDRINA-N
SMILES
NC1=C2N=CN([C@@H]3O[C@H](COP(O)(O)=O)[C@@H](O)[C@H]3O)C2=NC=N1
Identification (FTIR)
Conforms
Water Content (Karl Fischer Titration)
≤6%
Optical Rotation
-47.0° ± 2.0° (c=2 in 2% NaOH)
Appearance (Color)
White
Form
Powder

Description

a useful ligand determinant that facilitate the binding of APS reductase inhibitors and activates adenosine receptor agonists. 5?-AMP is an activator of a class of protein kinases known as AMP-activated protein kinase, an activator of phosphorylase b in the polymerization reaction for the synthesis of branched polysaccharides, a reagent in the synthesis of adenosine-5?-phosphoimidazolide, an inhibitor of endogenous NMN adenylyltransferase (NMNAT) activity that converts NMN to NAD+.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
a useful ligand determinant that facilitate the binding of APS reductase inhibitors and activates adenosine receptor agonists. 5′-AMP is an activator of a class of protein kinases known as AMP-activated protein kinase, an activator of phosphorylase b in the polymerization reaction for the synthesis of branched polysaccharides, a reagent in the synthesis of adenosine-5′-phosphoimidazolide, an inhibitor of endogenous NMN adenylyltransferase (NMNAT) activity that converts NMN to NAD+.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text