(1R,2R)-(-)-1,2-Diaminocyclohexane is a versatile ligand for the formation of metal complexes. Used in the synthesis of chiral tropocoronands which have potential utility in asymmetric catalysis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
(1R,2R)-(-)-1,2-Diaminocyclohexane is a versatile ligand for the formation of metal complexes. Used in the synthesis of chiral tropocoronands which have potential utility in asymmetric catalysis.
Solubility
Soluble in water.
Notes
Air sensitive. Store under inert gas. Store away from oxidizing agents, air, carbondioxide.
RUO – Research Use Only
General References:
- Catherine Bied; Joël J.E. Moreau; Michel Wong Chi Man. Chiral amino-urea derivatives of (1R,2R)-1,2-diaminocyclohexane as ligands in the ruthenium catalysed asymmetric reduction of aromatic ketones by hydride transfer. Tetrahedron: Asymmetry. 2001, 12, (2), 329-336.
- M. Kaik; J. Gawroński. Facile monoprotection of trans-1,2-diaminocyclohexane. Tetrahedron: Asymmetry. 2003, 14, (11), 1559-1563.