7,8-Dihydroxy-4-methylcoumarin, 97%, Thermo Scientific Chemicals
7,8-Dihydroxy-4-methylcoumarin, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

7,8-Dihydroxy-4-methylcoumarin, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mg
1 g
Catalog number L14069.ME
also known as L14069-ME
Price (USD)/ Each
36.80
-
Add to cart
Quantity:
500 mg
Request bulk or custom format
Price (USD)/ Each
36.80
Add to cart
7,8-Dihydroxy-4-methylcoumarin, 97%, Thermo Scientific Chemicals
Catalog numberL14069.ME
Price (USD)/ Each
36.80
-
Add to cart
Chemical Identifiers
CAS2107-77-9
IUPAC Name7,8-dihydroxy-4-methyl-2H-chromen-2-one
Molecular FormulaC10H8O4
InChI KeyNWQBYMPNIJXFNQ-UHFFFAOYSA-N
SMILESCC1=CC(=O)OC2=C(O)C(O)=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Melting Point (clear melt)238.0-244.0?C
Appearance (Color)White to pale brown
Assay (HPLC)≥96.0%
Can be used as a fluorescent sensor to monitor the consumption of a boronic acid in a Suzuki cross-coupling reaction. Use a long wave UV lamp.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only