Thermo Scientific Chemicals

(1S,2S)-(+)-1,2-Diaminocyclohexane, 98%, Thermo Scientific Chemicals

Catalog number: L14072.06
5 g, Each
Thermo Scientific Chemicals

(1S,2S)-(+)-1,2-Diaminocyclohexane, 98%, Thermo Scientific Chemicals

Catalog number: L14072.06
5 g, Each
Quantity
Catalog number: L14072.06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
21436-03-3
IUPAC Name
cyclohexane-1,2-diamine
Molecular Formula
C6H14N2
InChI Key
SSJXIUAHEKJCMH-UHFFFAOYNA-N
SMILES
NC1CCCCC1N
Optical Rotation
24.8 ± 0.1? (C=5 in 5N HCl)
Appearance (Color)
Colorless to white to pale cream to cream to yellow
Assay (GC)
≥97.5%
Form
Crystals or powder or crystalline powder or fused solid

Description

For synthesis of optically active products

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text