1-Aminopiperidine, 97%, Thermo Scientific Chemicals
1-Aminopiperidine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Aminopiperidine, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number L14078.06
also known as L14078-06
Price (USD)/ Each
58.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
58.00
Add to cart
1-Aminopiperidine, 97%, Thermo Scientific Chemicals
Catalog numberL14078.06
Price (USD)/ Each
58.00
-
Add to cart
Specifications
Chemical Name or Material1-Aminopiperidine
CAS2213-43-6
Health Hazard 1H226-H302+H312+H332-H314-H335
Health Hazard 2GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Health Hazard 3P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only