Thermo Scientific Chemicals

Tetra-n-pentylammonium iodide, 98%, Thermo Scientific Chemicals

Catalog number: L14088.14
25 g, Each
Thermo Scientific Chemicals

Tetra-n-pentylammonium iodide, 98%, Thermo Scientific Chemicals

Catalog number: L14088.14
25 g, Each
Quantity
Catalog number: L14088.14
also known as L14088-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2498-20-6
IUPAC Name
tetrapentylazanium iodide
Molecular Formula
C20H44IN
InChI Key
FBLZDUAOBOMSNZ-UHFFFAOYSA-M
SMILES
[I-].CCCCC[N+](CCCCC)(CCCCC)CCCCC
Form
Crystals or powder or crystalline powder
Assay (Titration ex Iodide)
≥97.5 to ≤102.5%
Appearance (Color)
White to cream or pale yellow
Melting Point (clear melt)
134.0-140.0?C

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text