Thermo Scientific Chemicals

7-Ethoxycoumarin, 99%, Thermo Scientific Chemicals

Catalog number: L14116.ME
500 mg, Each
Thermo Scientific Chemicals

7-Ethoxycoumarin, 99%, Thermo Scientific Chemicals

Catalog number: L14116.ME
500 mg, Each
Quantity
Catalog number: L14116.ME
also known as L14116-ME
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
31005-02-4
IUPAC Name
7-ethoxy-2H-chromen-2-one
Molecular Formula
C11H10O3
InChI Key
LIFAQMGORKPVDH-UHFFFAOYSA-N
SMILES
CCOC1=CC=C2C=CC(=O)OC2=C1
Appearance (Color)
White to cream
Form
Crystals or powder or crystalline powder
Assay (HPLC)
≥98.5%
Melting Point (clear melt)
86.5-92.5?C

Description

7-Ethoxycoumarin is a substrate for CYP2B6 used in fluorometric assays of microsomal monooxygenase activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
7-Ethoxycoumarin is a substrate for CYP2B6 used in fluorometric assays of microsomal monooxygenase activity.

Solubility
Soluble in hot methanol (very faint turbidity), 95% ethanol (50 mg/ml), DMF, and DMSO. Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text