delta-Nonanolactone, 98%, Thermo Scientific Chemicals
delta-Nonanolactone, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

delta-Nonanolactone, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number L14122.14
also known as L14122-14
Price (USD)/ Each
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS3301-94-8
IUPAC Name6-butyloxan-2-one
Molecular FormulaC9H16O2
InChI KeyPXRBWNLUQYZAAX-UHFFFAOYNA-N
SMILESCCCCC1CCCC(=O)O1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
Refractive Index1.4555-1.4595 @ 20?C
Appearance (Color)Clear colorless
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only