Thermo Scientific Chemicals

Methyl 3-bromopropionate, 97%, Thermo Scientific Chemicals

Catalog number: L14127.22
100 g, Each
Thermo Scientific Chemicals

Methyl 3-bromopropionate, 97%, Thermo Scientific Chemicals

Catalog number: L14127.22
100 g, Each
Quantity
Catalog number: L14127.22
also known as L14127-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3395-91-3
IUPAC Name
methyl 3-bromopropanoate
Molecular Formula
C4H7BrO2
InChI Key
KQEVIFKPZOGBMZ-UHFFFAOYSA-N
SMILES
COC(=O)CCBr
Appearance (Color)
Clear, coloorless to very pale yellow
Form
Liquid
Assay (GC)
>96.0%
Refractive Index
1.4555-1.4605 @ 20?c

Description

Methyl 3-bromopropionate was used in the synthesis of spiroanellated γ-lactones, pinacols and unprecedented 3-(1-hydroxycycloalkyl)-l-oxaspiro[n,m]alkan-2-ones. It can react with 2-Methyl-1H-benzoimidazole to get 1-(2-Methoxycarbonylethyl)-2-methylbenzimidazole.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 3-bromopropionate was used in the synthesis of spiroanellated γ-lactones, pinacols and unprecedented 3-(1-hydroxycycloalkyl)-l-oxaspiro[n,m]alkan-2-ones. It can react with 2-Methyl-1H-benzoimidazole to get 1-(2-Methoxycarbonylethyl)-2-methylbenzimidazole.

Solubility
Immiscible in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text