Amberlite XAD-4
Thermo Scientific Chemicals

Amberlite XAD-4

CAS: 37380-42-0
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number L14142.22
also known as L14142-22
Price (USD)
37.65
Online Exclusive
41.50
Save 3.85 (9%)
Each
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)
37.65
Online Exclusive
41.50
Save 3.85 (9%)
Each
Add to cart
Chemical Identifiers
CAS37380-42-0
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to yellow
FormBeads
CommentPonceau dye test: ≤1.0
Harmonic mean size0.490-0.690 mm
Moisture holding capacity54.0-60.0%
View more
Ask our AI about this Product
Polyaromatic adsorbent for small hydrophobic compounds, surfactants, pharmaceutical manufacturing, phenols, chlorinated organics and organic removal from aqueous food streams. It is impregnated with KCN or NaCN, has been used for the conversion of aromatic acyl chlorides to acyl cyanides and of formation of trialkylsilyl chlorides to the silyl cyanides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Polyaromatic adsorbent for small hydrophobic compounds, surfactants, pharmaceutical manufacturing, phenols, chlorinated organics and organic removal from aqueous food streams. It is impregnated with KCN or NaCN, has been used for the conversion of aromatic acyl chlorides to acyl cyanides and of formation of trialkylsilyl chlorides to the silyl cyanides.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.
RUO – Research Use Only

General References:

  1. Aimin Li; Quanxing Zhang; Jinlong Chen; Zhenghao Fei; Chao Long; Wanxing Li. Adsorption of phenolic compounds on Amberlite XAD-4 and its acetylated derivative MX-4. Reactive and Functional Polymers. 2001, 49, (3), 225-233.
  2. Attinti Ramesh; Kurakalva Rama Mohan; Kalluru Seshaiah. Preconcentration of trace metals on Amberlite XAD-4 resin coated with dithiocarbamates and determination by inductively coupled plasma-atomic emission spectrometry in saline matrices. Talanta. 2002, 57, (2), 243-252.
  3. Non-ionic polystyrene adsorbent, approximate surface area 750 m2/g.
  4. Impregnated with KCN or NaCN, has been used for the conversion of aromatic acyl chlorides to acyl cyanides: Bull. Chem. Soc. Jpn., 60, 1085 (1987), and of formation of trialkylsilyl chlorides to the silyl cyanides: Bull. Chem. Soc. Jpn., 60, 2289 (1987).
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link