Thermo Scientific Chemicals

Methyl (R)-(-)-3-hydroxybutyrate, 98%, Thermo Scientific Chemicals

Catalog number: L14151.14
25 g, Each
Thermo Scientific Chemicals

Methyl (R)-(-)-3-hydroxybutyrate, 98%, Thermo Scientific Chemicals

Catalog number: L14151.14
25 g, Each
Quantity
Catalog number: L14151.14
also known as L14151-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3976-69-0
IUPAC Name
methyl 3-hydroxybutanoate
Molecular Formula
C5H10O3
InChI Key
LDLDJEAVRNAEBW-UHFFFAOYNA-N
SMILES
COC(=O)CC(C)O
Optical Rotation
-49.0 ± 2.0? (C=1.3 in chloroform)
Appearance (Color)
Clear colorless
Form
Liquid
Assay (GC)
≥97.5%
Refractive Index
1.4195-1.4235 @ 20?C

Description

Can undergo aldol-type reactions with aldehydes with either syn- or anti-selectivity according to the conditions, for use in the synthesis of chiral β-lactams.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Can undergo aldol-type reactions with aldehydes with either syn- or anti-selectivity according to the conditions, for use in the synthesis of chiral ß-lactams.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text