Thermo Scientific Chemicals

6-Amino-1-hexanol, 97%, Thermo Scientific Chemicals

Catalog number: L14157.06
5 g, Each
Thermo Scientific Chemicals

6-Amino-1-hexanol, 97%, Thermo Scientific Chemicals

Catalog number: L14157.06
5 g, Each
Quantity
Catalog number: L14157.06
also known as L14157-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4048-33-3
IUPAC Name
6-aminohexan-1-ol
Molecular Formula
C6H15NO
InChI Key
SUTWPJHCRAITLU-UHFFFAOYSA-N
SMILES
NCCCCCCO
Form
Crystals or powder or crystalline powder or lumps or fused solid or liquid as melt
Assay (GC)
≥96.0%
Melting Point (clear melt)
51.0-61.0?C
Appearance (Color)
White to cream to yellow

Description

6-Amino-1-hexanol is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Amino-1-hexanol is used as pharmaceutical intermediate.

Solubility
Soluble in water.

Notes
Hygroscopic. Store away from oxidizing agents, water/ moisture.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text