Thermo Scientific Chemicals

5-Bromopyrimidine, 98%, Thermo Scientific Chemicals

Catalog number: L14172.22
100 g, Each
Thermo Scientific Chemicals

5-Bromopyrimidine, 98%, Thermo Scientific Chemicals

Catalog number: L14172.22
100 g, Each
Quantity
Catalog number: L14172.22
also known as L14172-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4595-59-9
IUPAC Name
5-bromopyrimidine
Molecular Formula
C4H3BrN2
InChI Key
GYCPLYCTMDTEPU-UHFFFAOYSA-N
SMILES
BrC1=CN=CN=C1
Assay (GC)
≥97.5%
Appearance (Color)
White to cream to pale brown
Form
Crystals or powder or crystalline powder or lumps or fused solid
Identification (FTIR)
Conforms

Description

5-Bromopyrimidine was used in the synthesis of N-heteroaryl substituted 9-arylcarbazolyl derivatives via palladium-catalyzed aerobic and ligand-free Suzuki reaction, (5-(phenylethynyl)pyrimidine) via microwave assisted organic synthesis (MAOS) Sonogashira protocol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Bromopyrimidine was used in the synthesis of N-heteroaryl substituted 9-arylcarbazolyl derivatives via palladium-catalyzed aerobic and ligand-free Suzuki reaction, (5-(phenylethynyl)pyrimidine) via microwave assisted organic synthesis (MAOS) Sonogashira protocol.

Solubility
Soluble in chloroform.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text